Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100000084.mm.1 |
NM_001166547.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000084.mm.1 |
NM_001166547.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000084.mm.1 |
NM_001166548.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000084.mm.1 |
NM_001166548.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000084.mm.1 |
NM_001166549.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000084.mm.1 |
NM_001166549.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000084.mm.1 |
NM_023743.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000084.mm.1 |
NM_023743.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000084.mm.1 |
TC1100000005.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100000082.mm.1 |
NM_001166547.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000082.mm.1 |
NM_001166547.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000082.mm.1 |
NM_001166548.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000082.mm.1 |
NM_001166548.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000082.mm.1 |
NM_001166549.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000082.mm.1 |
NM_001166549.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000082.mm.1 |
NM_023743.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000082.mm.1 |
NM_023743.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000082.mm.1 |
TC1100000005.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100000086.mm.1 |
NM_001166547.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000086.mm.1 |
NM_001166547.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000086.mm.1 |
NM_001166548.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000086.mm.1 |
NM_001166548.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000086.mm.1 |
NM_001166549.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000086.mm.1 |
NM_001166549.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000086.mm.1 |
NM_023743.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000086.mm.1 |
NM_023743.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000086.mm.1 |
TC1100000005.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100000088.mm.1 |
NM_001166547.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000088.mm.1 |
NM_001166547.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000088.mm.1 |
NM_001166548.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000088.mm.1 |
NM_001166548.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000088.mm.1 |
NM_001166549.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000088.mm.1 |
NM_001166549.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000088.mm.1 |
NM_023743.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000088.mm.1 |
NM_023743.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000088.mm.1 |
TC1100000005.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100000085.mm.1 |
NM_001166547.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000085.mm.1 |
NM_001166547.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000085.mm.1 |
NM_001166548.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000085.mm.1 |
NM_001166548.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000085.mm.1 |
NM_001166549.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000085.mm.1 |
NM_001166549.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000085.mm.1 |
NM_023743.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000085.mm.1 |
NM_023743.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000085.mm.1 |
TC1100000005.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100000087.mm.1 |
NM_001166547.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000087.mm.1 |
NM_001166547.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000087.mm.1 |
NM_001166548.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000087.mm.1 |
NM_001166548.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000087.mm.1 |
NM_001166549.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000087.mm.1 |
NM_001166549.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000087.mm.1 |
NM_023743.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000087.mm.1 |
NM_023743.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100000087.mm.1 |
TC1100000005.mm.1 |
MTA TC |