Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcs1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK012324.1 | - | 3 - 4 | 658 | 89 | |
AK078925.1 | - | 3 - 4 | 670 | 89 | ||
AK079385.1 | - | 3 - 4 | 666 | 89 | ||
BC019781.1 | - | 3 - 4 | 558 | 4036047 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |