Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zkscan6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK007308.1 | - | 2 - 3 | 571 | 79 | |
AK015125.1 | - | 2 - 3 | 547 | 79 | ||
BC043075.1 | - | 3 - 4 | 602 | 6404784 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |