Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cttnbp2nl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163332.1 | NP_001156804.1 | 5 - 6 | 92 | ||
NM_001163333.1 | NP_001156805.1 | 5 - 6 | 92 | |||
NM_030249.4 | NP_084525.1 | 5 - 6 | 92 | |||
GenBank mRNA | AK029901.1 | - | 5 - 6 | 92 | ||
AK030438.1 | - | 5 - 6 | 92 | |||
AK129359.1 | - | 5 - 6 | 626 | 92 | ||
BC003236.1 | - | 5 - 6 | 590 | 3498078 | 92 | |
BC006952.1 | - | 1 - 2 | 78 | 92 | ||
CV562515.1 | - | 5 - 6 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |