Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apobec3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB461446.1 | - | 5 - 6 | 695 | 129 | |
AK044394.1 | - | 5 - 6 | 764 | 129 | ||
AK154035.1 | - | 809 | 129 | |||
BC003314.1 | - | 5 - 6 | 786 | 3155422 | 129 | |
EU707568.1 | - | 5 - 6 | 695 | 129 | ||
EU707569.1 | - | 4 - 5 | 527 | 129 | ||
EU707571.1 | - | 4 - 5 | 527 | 129 | ||
GQ871501.1 | - | 5 - 6 | 692 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |