Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lta4h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008517.2 | NP_032543.2 | 1 - 2 | 277 | 87 | |
GenBank mRNA | AK032828.1 | - | 1 - 2 | 222 | 87 | |
AK134931.1 | - | 1 - 2 | 263 | 87 | ||
AK179027.1 | - | 1 - 2 | 47 | 87 | ||
BC021417.1 | - | 1 - 2 | 271 | 5004113 | 87 | |
BY793553.1 | - | 1 - 2 | 278 | 87 | ||
M63848.1 | - | 243 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Mus musculus Probe spans exons |