Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mccc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023644.4 | NP_076133.3 | 13 - 14 | 1706 | 127 | |
GenBank mRNA | AF310338.1 | - | 13 - 14 | 1627 | 127 | |
AK007782.1 | - | 13 - 14 | 1659 | 127 | ||
AK031072.1 | - | 13 - 14 | 1667 | 127 | ||
AK032203.1 | - | 13 - 14 | 1649 | 127 | ||
AK168589.1 | - | 13 - 14 | 1595 | 127 | ||
AK201441.1 | - | 1 - 2 | 150 | 127 | ||
BC021382.1 | - | 13 - 14 | 1670 | 5042993 | 127 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |