Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcoln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053177.1 | NP_444407.1 | 4 - 5 | 653 | 111 | |
GenBank mRNA | AF302009.1 | - | 4 - 5 | 650 | 111 | |
AK028385.1 | - | 4 - 5 | 630 | 111 | ||
AK034471.1 | - | 4 - 5 | 664 | 111 | ||
AK038594.1 | - | 4 - 5 | 666 | 111 | ||
AK049606.1 | - | 4 - 5 | 654 | 111 | ||
BC005651.1 | - | 4 - 5 | 653 | 3257440 | 111 | |
BC020377.1 | - | 3 - 4 | 503 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel Mammalian Gene Collection Mus musculus Probe spans exons |