Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ggt7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144786.2 | NP_659035.2 | 13 - 14 | 1770 | 69 | |
GenBank mRNA | AF332053.1 | - | 1 - 2 | 68 | 69 | |
AF332054.1 | - | 1 - 2 | 68 | 69 | ||
AK032051.1 | - | 13 - 14 | 1768 | 69 | ||
AK131175.1 | - | 10 - 11 | 3793 | 69 | ||
BC005772.1 | - | 13 - 14 | 1751 | 3602314 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Mus musculus Probe spans exons |