Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mib1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144860.2 | NP_659109.2 | 14 - 15 | 2735 | 95 | |
GenBank mRNA | AK053035.1 | - | 7 - 8 | 958 | 95 | |
AK080847.1 | - | 8 - 9 | 1011 | 95 | ||
AY147848.1 | - | 14 - 15 | 2234 | 95 | ||
AY149907.1 | - | 14 - 15 | 2055 | 95 | ||
AY245539.1 | - | 14 - 15 | 2735 | 95 | ||
AY974091.1 | - | 14 - 15 | 2028 | 95 | ||
BC011287.1 | - | 3 - 4 | 332 | 95 | ||
BC083072.1 | - | 4 - 5 | 413 | 95 | ||
BC131662.1 | - | 15 - 16 | 2201 | 40110696 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |