Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cox15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144874.4 | NP_659123.2 | 1 - 2 | 182 | 119 | |
GenBank mRNA | AK048099.1 | - | 1 - 2 | 164 | 119 | |
AK051792.1 | - | 1 - 2 | 80 | 119 | ||
AK141631.1 | - | 1 - 2 | 114 | 119 | ||
BC011509.1 | - | 1 - 2 | 142 | 3981950 | 119 | |
BC021498.1 | - | 1 - 2 | 102 | 119 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |