Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zgpat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048148.1 | NP_001041613.1 | 4 - 5 | 1235 | 63 | |
NM_144894.3 | NP_659143.1 | 5 - 6 | 1459 | 63 | ||
GenBank mRNA | AK041491.1 | - | 5 - 6 | 1463 | 63 | |
AK049764.1 | - | 4 - 5 | 1234 | 63 | ||
AK052236.1 | - | 4 - 5 | 1133 | 63 | ||
AK159801.1 | - | 4 - 5 | 1254 | 63 | ||
BC021513.1 | - | 4 - 5 | 1233 | 5351756 | 63 | |
BC027218.1 | - | 4 - 5 | 1142 | 3158198 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |