Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042655.1 | NP_001036120.1 | 11 - 12 | 1460 | 96 | |
GenBank mRNA | AK039566.1 | - | 11 - 12 | 1275 | 96 | |
AK046883.1 | - | 11 - 12 | 1294 | 96 | ||
AK083153.1 | - | 11 - 12 | 1278 | 96 | ||
AK087754.1 | - | 1286 | 96 | |||
AK136705.1 | - | 11 - 12 | 1285 | 96 | ||
AK149917.1 | - | 11 - 12 | 1272 | 96 | ||
AK153732.1 | - | 11 - 12 | 1272 | 96 | ||
AK170821.1 | - | 11 - 12 | 1272 | 96 | ||
BC017607.1 | - | 11 - 12 | 1460 | 4507589 | 96 | |
BC035449.1 | - | 11 - 12 | 1259 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |