Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700015645.mm.1 |
TC0700004626.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700015645.mm.1 |
NM_001252566.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700015645.mm.1 |
NM_001252566.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700015645.mm.1 |
NM_001252567.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700015645.mm.1 |
NM_001252567.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700015645.mm.1 |
NM_144926.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700015645.mm.1 |
NM_144926.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700015638.mm.1 |
TC0700004626.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700015638.mm.1 |
NM_001252566.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700015638.mm.1 |
NM_001252566.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700015638.mm.1 |
NM_001252567.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700015638.mm.1 |
NM_001252567.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700015638.mm.1 |
NM_144926.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700015638.mm.1 |
NM_144926.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700015644.mm.1 |
TC0700004626.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700015644.mm.1 |
NM_001252566.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700015644.mm.1 |
NM_001252566.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700015644.mm.1 |
NM_001252567.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700015644.mm.1 |
NM_001252567.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700015644.mm.1 |
NM_144926.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700015644.mm.1 |
NM_144926.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700039158.mm.1 |
NM_001252566.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700039158.mm.1 |
NM_001252566.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700039158.mm.1 |
NM_001252567.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700039158.mm.1 |
NM_001252567.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700039158.mm.1 |
NM_144926.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700039158.mm.1 |
NM_144926.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700039158.mm.1 |
TC0700004626.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700015640.mm.1 |
TC0700004626.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700015640.mm.1 |
NM_001252566.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700015640.mm.1 |
NM_001252566.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700015640.mm.1 |
NM_001252567.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700015640.mm.1 |
NM_001252567.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700015640.mm.1 |
NM_144926.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700015640.mm.1 |
NM_144926.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700015641.mm.1 |
TC0700004626.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700015641.mm.1 |
NM_001252566.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700015641.mm.1 |
NM_001252566.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700015641.mm.1 |
NM_001252567.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700015641.mm.1 |
NM_001252567.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700015641.mm.1 |
NM_144926.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700015641.mm.1 |
NM_144926.5 |
Refseq |