Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhh3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146030.2 | NP_666142.1 | 9 - 10 | 1840 | 96 | |
GenBank mRNA | AK052371.1 | - | 1 - 2 | 40 | 96 | |
BC020025.1 | - | 9 - 10 | 1840 | 3490992 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |