Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ipo13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146152.3 | NP_666264.1 | 16 - 17 | 3093 | 61 | |
GenBank mRNA | AK052257.1 | - | 16 - 17 | 3094 | 61 | |
AK129200.1 | - | 16 - 17 | 3055 | 61 | ||
AK149978.1 | - | 16 - 17 | 3076 | 61 | ||
AK192686.1 | - | 1 - 2 | 59 | 61 | ||
BC031823.1 | - | 16 - 17 | 2492 | 4188768 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |