Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700014287.mm.1 |
TC0700001710.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700014287.mm.1 |
NM_001177978.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700014287.mm.1 |
NM_001177978.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700014287.mm.1 |
NM_146197.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700014287.mm.1 |
NM_146197.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700014291.mm.1 |
TC0700001710.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700014291.mm.1 |
NM_001177977.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700014291.mm.1 |
NM_001177977.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700014291.mm.1 |
NM_001177978.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700014291.mm.1 |
NM_001177978.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700014291.mm.1 |
NM_146197.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700014291.mm.1 |
NM_146197.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700014282.mm.1 |
TC0700001710.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700014282.mm.1 |
NM_001177977.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700014282.mm.1 |
NM_001177977.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700014290.mm.1 |
TC0700001710.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700014290.mm.1 |
NM_001177977.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700014290.mm.1 |
NM_001177977.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700014290.mm.1 |
NM_001177978.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700014290.mm.1 |
NM_001177978.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700014290.mm.1 |
NM_146197.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700014290.mm.1 |
NM_146197.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700014289.mm.1 |
TC0700001710.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700014289.mm.1 |
NM_001177977.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700014289.mm.1 |
NM_001177977.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700014289.mm.1 |
NM_001177978.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700014289.mm.1 |
NM_001177978.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700014289.mm.1 |
NM_146197.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700014289.mm.1 |
NM_146197.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700014283.mm.1 |
TC0700001710.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700014283.mm.1 |
NM_001177977.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700014283.mm.1 |
NM_001177977.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700014288.mm.1 |
TC0700001710.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700014288.mm.1 |
NM_001177978.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700014288.mm.1 |
NM_001177978.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700014288.mm.1 |
NM_146197.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700014288.mm.1 |
NM_146197.4 |
Refseq |