Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mdm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162904.1 | NP_001156376.1 | 6 - 7 | 1063 | 84 | |
NM_001162905.1 | NP_001156377.1 | 5 - 6 | 928 | 84 | ||
NM_010785.2 | NP_034915.2 | 6 - 7 | 1063 | 84 | ||
NM_148922.3 | NP_683724.2 | 6 - 7 | 1063 | 84 | ||
GenBank mRNA | AK004789.2 | - | 6 - 7 | 1018 | 84 | |
AK011769.1 | - | 6 - 7 | 1018 | 84 | ||
BC052680.1 | - | 6 - 7 | 1013 | 84 | ||
BC137608.1 | - | 6 - 7 | 991 | 8860627 | 84 | |
BC144777.1 | - | 6 - 7 | 991 | 9053325 | 84 | |
EU490427.1 | - | 6 - 7 | 905 | 84 | ||
EU490428.1 | - | 5 - 6 | 770 | 84 | ||
M20823.1 | - | 6 - 7 | 926 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |