Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Angpt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007426.4 | NP_031452.2 | 2 - 3 | 732 | 67 | |
GenBank mRNA | AF004326.1 | - | 2 - 3 | 658 | 67 | |
AK019860.1 | - | 2 - 3 | 721 | 67 | ||
AK048622.1 | - | 2 - 3 | 689 | 67 | ||
AK156132.1 | - | 2 - 3 | 703 | 67 | ||
BC027216.1 | - | 2 - 3 | 685 | 3494566 | 67 | |
BC138309.1 | - | 2 - 3 | 626 | 8861329 | 67 | |
BC138313.1 | - | 2 - 3 | 626 | 8861333 | 67 | |
CN539462.1 | - | 2 - 3 | 733 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |