Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cry2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009963.4 | NP_034093.1 | 6 - 7 | 76 | ||
GenBank mRNA | AB003433.1 | - | 6 - 7 | 76 | ||
AF156987.1 | - | 6 - 7 | 76 | |||
AK041696.1 | - | 6 - 7 | 76 | |||
AK133781.1 | - | 6 - 7 | 76 | |||
AK172994.1 | - | 6 - 7 | 76 | |||
BC054794.1 | - | 6 - 7 | 6834104 | 76 | ||
BC066799.1 | - | 6 - 7 | 30103169 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |