Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172443.3 | NP_766031.2 | 7 - 8 | 1564 | 74 | |
GenBank mRNA | AK047555.1 | - | 7 - 8 | 1565 | 74 | |
BC057634.1 | - | 7 - 8 | 1523 | 2866041 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |