Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Def6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027185.3 | NP_081461.2 | 8 - 9 | 1446 | 93 | |
GenBank mRNA | AF329497.1 | - | 8 - 9 | 1381 | 93 | |
AK038050.1 | - | 8 - 9 | 1417 | 93 | ||
AK088460.1 | - | 8 - 9 | 1395 | 93 | ||
AY241695.1 | - | 8 - 9 | 1381 | 93 | ||
AY278770.1 | - | 2 - 3 | 262 | 93 | ||
BC120500.1 | - | 8 - 9 | 1446 | 40129423 | 93 | |
BC131953.1 | - | 8 - 9 | 1446 | 40130230 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |