Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Golm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035122.2 | NP_001030294.1 | 7 - 8 | 821 | 125 | |
NM_027307.4 | NP_081583.3 | 8 - 9 | 957 | 125 | ||
GenBank mRNA | AK009072.1 | - | 6 - 7 | 628 | 125 | |
AK133865.1 | - | 7 - 8 | 823 | 125 | ||
AK167646.1 | - | 7 - 8 | 798 | 125 | ||
BC011152.1 | - | 7 - 8 | 782 | 4188434 | 125 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |