Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dmrtc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027732.2 | NP_082008.1 | 6 - 7 | 880 | 85 | |
GenBank mRNA | AF542046.1 | - | 6 - 7 | 877 | 85 | |
AK017020.1 | - | 6 - 7 | 878 | 85 | ||
BC100434.1 | - | 6 - 7 | 880 | 30918218 | 85 | |
BC138203.1 | - | 6 - 7 | 811 | 8861222 | 85 | |
BC145988.1 | - | 6 - 7 | 811 | 40131291 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |