Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcor | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168321.1 | NP_001161793.1 | 5 - 6 | 3310 | 116 | |
NM_029510.3 | NP_083786.2 | 6 - 7 | 4033 | 116 | ||
NM_175044.3 | NP_778209.2 | 5 - 6 | 3979 | 116 | ||
NM_175045.3 | NP_778210.2 | 6 - 7 | 4033 | 116 | ||
NM_175046.3 | NP_778211.2 | 5 - 6 | 3979 | 116 | ||
GenBank mRNA | AK129398.1 | - | 5 - 6 | 3310 | 116 | |
AY161170.1 | - | 6 - 7 | 3348 | 116 | ||
AY161171.1 | - | 5 - 6 | 3294 | 116 | ||
AY161172.1 | - | 6 - 7 | 3348 | 116 | ||
AY161173.1 | - | 5 - 6 | 3294 | 116 | ||
BC058656.1 | - | 6 - 7 | 3534 | 6412868 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |