Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Evi5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039578.3 | NP_001034667.3 | 6 - 7 | 998 | 74 | |
GenBank mRNA | AK045240.1 | - | 6 - 7 | 999 | 74 | |
BC042653.1 | - | 6 - 7 | 978 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |