Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sbsn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172205.3 | NP_757342.2 | 1 - 2 | 2001 | 56 | |
GenBank mRNA | AA791703.1 | - | 1 - 2 | 245 | 56 | |
AI507307.1 | - | 3 - 4 | 56 | |||
AY115494.1 | - | 1 - 2 | 1994 | 56 | ||
BC139385.1 | - | 1 - 2 | 1993 | 8862407 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |