Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172338.2 | NP_758841.1 | 6 - 7 | 933 | 92 | |
GenBank mRNA | AK028692.1 | - | 6 - 7 | 938 | 92 | |
AK051048.1 | - | 6 - 7 | 934 | 92 | ||
AK122407.1 | - | 1180 | 92 | |||
AK154946.1 | - | 954 | 92 | |||
AK164023.1 | - | 6 - 7 | 949 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |