Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itih5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172471.2 | NP_766059.1 | 3 - 4 | 420 | 77 | |
GenBank mRNA | AK077381.1 | - | 343 | 77 | ||
AK087428.1 | - | 3 - 4 | 348 | 77 | ||
AK134707.1 | - | 3 - 4 | 348 | 77 | ||
AK140756.1 | - | 3 - 4 | 421 | 77 | ||
BB595829.2 | - | 3 - 4 | 421 | 77 | ||
BC043314.1 | - | 3 - 4 | 363 | 5150581 | 77 | |
BC062196.1 | - | 3 - 4 | 404 | 3490706 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |