Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem71 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172514.3 | NP_766102.1 | 6 - 7 | 1009 | 80 | |
GenBank mRNA | AK088711.1 | - | 6 - 7 | 1010 | 80 | |
AK170268.1 | - | 6 - 7 | 1006 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |