Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mppe1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172630.2 | NP_766218.1 | 8 - 9 | 1387 | 61 | |
GenBank mRNA | AK041262.1 | - | 8 - 9 | 1388 | 61 | |
AK154265.1 | - | 8 - 9 | 1241 | 61 | ||
BC045146.1 | - | 8 - 9 | 1010 | 6306245 | 61 | |
BC086456.1 | - | 8 - 9 | 1057 | 4949311 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |