Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172704.3 | NP_766292.2 | 1 - 2 | 107 | 81 | |
GenBank mRNA | AK053156.1 | - | 1 - 2 | 89 | 81 | |
AK089983.1 | - | 1 - 2 | 99 | 81 | ||
AK156793.1 | - | 1 - 2 | 89 | 81 | ||
AK171781.1 | - | 1 - 2 | 88 | 81 | ||
AK172542.1 | - | 1 - 2 | 88 | 81 | ||
BC075619.1 | - | 1 - 2 | 83 | 81 | ||
BC079852.1 | - | 1 - 2 | 77 | 81 | ||
BC085257.1 | - | 1 - 2 | 81 | 5414963 | 81 | |
BY784439.1 | - | 1 - 2 | 117 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |