Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500024285.mm.1 |
TC0500002754.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500024285.mm.1 |
NM_172713.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024285.mm.1 |
NM_172713.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024287.mm.1 |
TC0500002754.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500024287.mm.1 |
NM_172713.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024287.mm.1 |
NM_172713.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024284.mm.1 |
TC0500002754.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500024284.mm.1 |
NM_172713.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024284.mm.1 |
NM_172713.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024292.mm.1 |
TC0500002754.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500024292.mm.1 |
NM_172713.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024292.mm.1 |
NM_172713.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024298.mm.1 |
TC0500002754.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500024298.mm.1 |
NM_172713.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024298.mm.1 |
NM_172713.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024283.mm.1 |
TC0500002754.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500024283.mm.1 |
NM_172713.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024283.mm.1 |
NM_172713.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024294.mm.1 |
TC0500002754.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500024294.mm.1 |
NM_172713.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024294.mm.1 |
NM_172713.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024297.mm.1 |
TC0500002754.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500024297.mm.1 |
NM_172713.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024297.mm.1 |
NM_172713.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024289.mm.1 |
TC0500002754.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500024289.mm.1 |
NM_172713.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024289.mm.1 |
NM_172713.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024291.mm.1 |
TC0500002754.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500024291.mm.1 |
NM_172713.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024291.mm.1 |
NM_172713.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024299.mm.1 |
TC0500002754.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500024299.mm.1 |
NM_172713.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024299.mm.1 |
NM_172713.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024290.mm.1 |
TC0500002754.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500024290.mm.1 |
NM_172713.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024290.mm.1 |
NM_172713.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024293.mm.1 |
TC0500002754.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500024293.mm.1 |
NM_172713.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024293.mm.1 |
NM_172713.2 |
Refseq |