Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sdad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172713.2 | NP_766301.1 | 13 - 14 | 1250 | 69 | |
GenBank mRNA | AK029881.1 | - | 13 - 14 | 1252 | 69 | |
AK160646.1 | - | 13 - 14 | 1118 | 69 | ||
BC042708.1 | - | 13 - 14 | 1200 | 5024980 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |