Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dera | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172733.1 | NP_766321.1 | 1 - 2 | 61 | 80 | |
GenBank mRNA | AK012558.1 | - | 1 - 2 | 97 | 80 | |
AK030961.1 | - | 1 - 2 | 93 | 80 | ||
AK034347.1 | - | 63 | 80 | |||
AK075963.1 | - | 1 - 2 | 86 | 80 | ||
AK083298.1 | - | 1 - 2 | 61 | 80 | ||
AK145820.1 | - | 1 - 2 | 97 | 80 | ||
AK150273.1 | - | 1 - 2 | 87 | 80 | ||
AK150712.1 | - | 1 - 2 | 57 | 80 | ||
AK152462.1 | - | 1 - 2 | 65 | 80 | ||
AK168204.1 | - | 1 - 2 | 97 | 80 | ||
AK169418.1 | - | 1 - 2 | 81 | 80 | ||
BC016218.1 | - | 1 - 2 | 67 | 4484163 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |