Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173413.3 | NP_775589.1 | 2 - 3 | 258 | 86 | |
GenBank mRNA | AB232601.1 | - | 2 - 3 | 186 | 86 | |
AK031251.1 | - | 2 - 3 | 205 | 86 | ||
AK051218.1 | - | 2 - 3 | 208 | 86 | ||
AK084650.1 | - | 2 - 3 | 208 | 86 | ||
AK155621.1 | - | 2 - 3 | 208 | 86 | ||
BC059208.1 | - | 2 - 3 | 240 | 6405462 | 86 | |
BC157974.1 | - | 2 - 3 | 206 | 9007296 | 86 | |
BY322741.1 | - | 2 - 3 | 259 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |