Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Card11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175362.2 | NP_780571.2 | 3 - 4 | 541 | 62 | |
GenBank mRNA | AK131134.1 | - | 3 - 4 | 541 | 62 | |
AY135367.1 | - | 3 - 4 | 437 | 62 | ||
BC096592.1 | - | 3 - 4 | 527 | 5318165 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons |