Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lsr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164184.1 | NP_001157656.1 | 7 - 8 | 1356 | 62 | |
NM_001164185.1 | NP_001157657.1 | 6 - 7 | 1209 | 62 | ||
NM_017405.2 | NP_059101.1 | 8 - 9 | 1413 | 62 | ||
GenBank mRNA | AK146807.1 | - | 7 - 8 | 1072 | 62 | |
AK167463.1 | - | 6 - 7 | 1148 | 62 | ||
BC004672.1 | - | 8 - 9 | 1159 | 3500090 | 62 | |
U49507.1 | - | 8 - 9 | 1304 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |