Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgs9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001165934.1 | NP_001159406.1 | 14 - 15 | 1241 | 88 | |
NM_011268.2 | NP_035398.2 | 14 - 15 | 1241 | 88 | ||
GenBank mRNA | AF011358.1 | - | 14 - 15 | 1126 | 88 | |
AF125046.1 | - | 14 - 15 | 1197 | 88 | ||
AK138159.1 | - | 14 - 15 | 1151 | 88 | ||
BC129898.1 | - | 6 - 7 | 519 | 40111530 | 88 | |
BC129899.1 | - | 14 - 15 | 1107 | 40111531 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |