Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013770.2 | NP_038798.2 | 6 - 7 | 609 | 69 | |
GenBank mRNA | AF188712.1 | - | 6 - 7 | 580 | 69 | |
AK008038.1 | - | 6 - 7 | 579 | 69 | ||
AK050282.1 | - | 6 - 7 | 563 | 69 | ||
AK083831.1 | - | 6 - 7 | 587 | 69 | ||
AK151645.1 | - | 6 - 7 | 609 | 69 | ||
BC003222.1 | - | 6 - 7 | 527 | 3482316 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |