Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013900.2 | NP_038928.1 | 2 - 3 | 318 | 57 | |
GenBank mRNA | AB024336.1 | - | 2 - 3 | 318 | 57 | |
AK036444.1 | - | 2 - 3 | 253 | 57 | ||
BC040347.1 | - | 2 - 3 | 241 | 3672916 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |