Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sart1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016882.3 | NP_058578.3 | 15 - 16 | 2041 | 58 | |
GenBank mRNA | AB014721.1 | - | 15 - 16 | 1999 | 58 | |
AF129931.1 | - | 15 - 16 | 2041 | 58 | ||
AK088341.1 | - | 15 - 16 | 2002 | 58 | ||
BC028823.1 | - | 15 - 16 | 2003 | 5097832 | 58 | |
BC051394.1 | - | 15 - 16 | 2003 | 5097832 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |