Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P21152827 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P21152831 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P21152830 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P256282 |
AK009104.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P256282 |
AK046859.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P256282 |
AK008835.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P256282 |
BC002223.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P256282 |
NM_024209.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P256282 |
AK009104.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P256282 |
AK046859.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P256282 |
AK008835.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P256282 |
BC002223.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P256282 |
NM_024209.2 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
428057 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
481675 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
518583 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
54064 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
451188 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
835130 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
524800 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
703964 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
212977 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
584506 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
642655 |
AK083130.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
642655 |
AK148688.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
642655 |
BC072641.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
642655 |
NM_020610.1 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
692441 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
640389 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
43763 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
127130 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
73319 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
849486 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
379018 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
164014 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
505498 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
50176 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
83442 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
84950 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P163928 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P108148 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P159291 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P256282 |
AK009104.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P256282 |
AK046859.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P256282 |
AK008835.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P256282 |
BC002223.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P256282 |
NM_024209.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P256282 |
AK009104.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P256282 |
AK046859.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P256282 |
AK008835.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P256282 |
BC002223.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P256282 |
NM_024209.2 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21152831 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21152821 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21152839 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21152841 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21152820 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21152823 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21152847 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21152837 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21152843 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21152838 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21152833 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21152834 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21152835 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21152849 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21152842 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21152846 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21152850 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21152845 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21152819 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21152822 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21152852 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21152832 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P00349454 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P163928 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P159291 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P108148 |
- |
- |
Whole Mouse Genome G4122A |
A_52_P243109 |
AK009104.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P243109 |
BC002223.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P243109 |
NM_024209.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P243109 |
AK002764.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P256282 |
AK009104.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P256282 |
AK046859.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P256282 |
AK008835.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P256282 |
BC002223.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P256282 |
NM_024209.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P256282 |
AK009104.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P256282 |
AK046859.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P256282 |
AK008835.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P256282 |
BC002223.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P256282 |
NM_024209.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P204440 |
AK046859.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P133534 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P163928 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P108148 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P04492484 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P159291 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P01764411 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P01666729 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P01666725 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P21152830 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P21152831 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P21152827 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152792 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152790 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152819 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152677 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152650 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152768 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152747 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152843 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152787 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152673 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152654 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152706 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152644 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152674 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152795 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152729 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152693 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152625 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152759 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152667 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152665 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152616 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152614 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152701 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152809 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152846 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152850 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152649 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152833 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152783 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152796 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152627 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152732 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152704 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152703 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152660 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152698 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152744 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152845 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152839 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152658 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152784 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152631 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152714 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152765 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152769 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152717 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152728 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152734 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152841 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152699 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152794 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152641 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152823 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152664 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152847 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152697 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152691 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152681 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152849 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152737 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152837 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152842 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152743 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152806 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152656 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152726 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152822 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152705 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152716 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152680 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152776 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152821 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152800 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152671 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152669 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152760 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152788 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152700 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152637 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152672 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152750 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152754 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152708 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152779 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152620 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152682 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152736 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152712 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152785 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152647 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152621 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152651 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152668 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152838 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152774 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152802 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152832 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152724 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152804 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152626 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152653 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152685 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152781 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152633 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152639 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152695 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152820 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152688 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152742 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152789 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152771 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152767 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152683 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21152692 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21152823 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21152843 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01315963 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21152796 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21152795 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21152836 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21152695 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21152831 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21152847 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21152806 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21152693 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21152842 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21152697 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01967702 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01718305 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01655811 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21152821 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21152832 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21152852 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21152833 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01289060 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01543683 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21152850 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21152792 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21152817 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21152839 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21152820 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21152838 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21152691 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01354171 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21152835 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01638454 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21152800 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01655804 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21152688 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01655820 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21152802 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21152692 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21152837 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01638452 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01655813 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21152813 |
- |
- |