Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam20b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145413.4 | NP_663388.1 | 5 - 6 | 913 | 62 | |
GenBank mRNA | AK049705.1 | - | 4 - 5 | 715 | 62 | |
AK129154.1 | - | 3 - 4 | 322 | 62 | ||
AK132149.1 | - | 5 - 6 | 914 | 62 | ||
BC019381.1 | - | 5 - 6 | 932 | 4206622 | 62 | |
BC023737.1 | - | 6 - 7 | 1129 | 5352660 | 62 | |
BC024412.1 | - | 4 - 5 | 863 | 4971483 | 62 | |
BC031473.1 | - | 5 - 6 | 1025 | 3990487 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |