Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam73a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162375.1 | NP_001155847.1 | 3 - 4 | 650 | 89 | |
NM_174868.4 | NP_777357.2 | 3 - 4 | 650 | 89 | ||
GenBank mRNA | AK044739.1 | - | 3 - 4 | 626 | 89 | |
AK045347.1 | - | 3 - 4 | 649 | 89 | ||
AK083909.1 | - | 2 - 3 | 481 | 89 | ||
AK085017.1 | - | 3 - 4 | 650 | 89 | ||
AK143366.1 | - | 3 - 4 | 649 | 89 | ||
AK166051.1 | - | 3 - 4 | 601 | 89 | ||
BC059928.1 | - | 3 - 4 | 616 | 89 | ||
BC098205.1 | - | 3 - 4 | 633 | 6409928 | 89 | |
BC138328.1 | - | 3 - 4 | 597 | 8861348 | 89 | |
BC138329.1 | - | 3 - 4 | 597 | 8861349 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |