Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ahnak | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039959.2 | NP_001035048.1 | 2 - 3 | 260 | 72 | |
NM_001286518.1 | NP_001273447.1 | 2 - 3 | 260 | 72 | ||
NM_009643.2 | NP_033773.1 | 2 - 3 | 260 | 72 | ||
NR_104460.1 | 2 - 3 | 260 | 72 | |||
GenBank mRNA | AK009866.1 | - | 2 - 3 | 283 | 72 | |
AK045184.1 | - | 2 - 3 | 261 | 72 | ||
BC028439.1 | - | 1 - 2 | 125 | 72 | ||
CB321491.1 | - | 2 - 3 | 243 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons |