Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fem1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK031720.1 | - | 2556 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus 1700 Inventoried |