Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptk7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175168.4 | NP_780377.1 | 8 - 9 | 1515 | 80 | |
GenBank mRNA | AF531873.1 | - | 8 - 9 | 1365 | 80 | |
AK053044.1 | - | 8 - 9 | 1448 | 80 | ||
AK163135.1 | - | 8 - 9 | 1448 | 80 | ||
BC076578.1 | - | 8 - 9 | 1515 | 30619280 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |