Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800023268.mm.1 |
NM_175185.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800023268.mm.1 |
NM_175185.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800023268.mm.1 |
TC0800003024.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800023267.mm.1 |
NM_175185.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800023267.mm.1 |
NM_175185.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800023267.mm.1 |
TC0800003024.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800023270.mm.1 |
NM_175185.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800023270.mm.1 |
NM_175185.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800023270.mm.1 |
TC0800003024.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800023271.mm.1 |
NM_175185.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800023271.mm.1 |
NM_175185.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800023271.mm.1 |
TC0800003024.mm.1 |
MTA TC |