Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec24a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290785.1 | NP_001277714.1 | 3 - 4 | 1388 | 102 | |
NM_175255.3 | NP_780464.2 | 3 - 4 | 1388 | 102 | ||
GenBank mRNA | AK029139.1 | - | 3 - 4 | 1141 | 102 | |
AK155182.1 | - | 3 - 4 | 1174 | 102 | ||
AK162795.1 | - | 3 - 4 | 993 | 102 | ||
AK163254.1 | - | 3 - 4 | 1717 | 103 | ||
AK163998.1 | - | 3 - 4 | 985 | 102 | ||
BC117905.1 | - | 3 - 4 | 652 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |